Order Entry
Mexico
Orders LinkContactUsLinkComponent
Digital Probe Thermometer

Digital Probe Thermometer

–50…+200 °C (–58…+392 °F), ±0.5 ˚C from –20...70 ˚C, ±1˚C from 70...120 ˚C, otherwise ±2 ˚C, 0.1° (-19.9 to 199°), 12.7 cm (5")
Catalog # 470204-692
Supplier:   BEL-ART PRODUCTS SE
Supplier #:  B60900-0400_SE
Digital Probe Thermometer
Catalog # 470204-692
Digital Probe Thermometer
Digital Probe Thermometer

–50…+200 °C (–58…+392 °F), ±0.5 ˚C from –20...70 ˚C, ±1˚C from 70...120 ˚C, otherwise ±2 ˚C, 0.1° (-19.9 to 199°), 12.7 cm (5")

Price
$1,920.00
Log in for availability
Each (1UN)
Product Details & Documents

Easy to Use.

  • Features a stainless steel probe
  • Has max/min temperature memory and data hold function
  • Easily switches from Fahrenheit to Celsius scale

Digital Probe Thermometer comes with a magnetic attachment, 5" probe, 59" probe cord, and one 1.5 V AAA battery. Size: 21/2"W x 33/4"H x 3/4"D.

Specifications
  • Resolution:
    0.1° (-19.9 to 199°)
  • Length:
    12.7 cm (5")
  • Accuracy:
    ±0.5 ˚C from –20...70 ˚C, ±1˚C from 70...120 ˚C, otherwise ±2 ˚C
  • Range:
    –50…+200 °C (–58…+392 °F)