Order Entry
Mexico
ContactUsLinkComponent
Diatoms Of North America
Diatoms Of North America
Catalog # 470022-604
Supplier:  MAD RIVER PRESS
undefined
Diatoms Of North America
Catalog # 470022-604
Supplier:  MAD RIVER PRESS
Supplier Number:  0916422151

Specifications

  • Description:
    Diatoms Of North America
  • Length:
    15 cm
  • Cat. no.:
    470022-604
  • No. of Pages:
    119
  • Width:
    22 cm
  • Supplier No.:
    0916422151

Specifications

About this item

William C. Vinyard

You’ll refer to this handy guide over and over for its detailed, illustrated key of freshwater and marine diatoms, and complete description of 110 genera, to better verify the determination at this taxonomic level. One hundred eighty forms in all are illustrated and identified.