Order Entry
Mexico
ContactUsLinkComponent
Dental Probe And Seeker
Dental Probe And Seeker
Catalog # 470007-562
undefined
Dental Probe And Seeker
Catalog # 470007-562
Supplier Number:  140955
Especificaciones
  • Description:
    Dental Probe And Seeker
  • Length:
    152 mm (6")
  • Material:
    Stainless steel
  • Cat. no.:
    470007-562
  • Supplier No.:
    140955
Especificaciones
Acerca de este artículo
The curved end has a knife edge for easy separation of tissue from bone, while the other end comes to a tapered, sharp point great for tracing arteries, or teasing muscle.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.