Culture Flasks
Supplier: CORNING CA
Jump to:
About this item
Specially Coated Tissue Culture Flasks.
- Nunclon surface treatment promotes cell attachment and growth
- Easy filling and access for pipets
- Stackable, space saving design
Cell adherence is discouraged from the untreated sidewalls, neck and top panel.
1 Options Available Below
Product Details & Documents
Specially Coated Tissue Culture Flasks.
- Nunclon surface treatment promotes cell attachment and growth
- Easy filling and access for pipets
- Stackable, space saving design
Cell adherence is discouraged from the untreated sidewalls, neck and top panel.
Documents
Certificates
Search by Lot NumberThermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Product Family Options
Product Information
- AvailabilityPrice
- 470174-45050 mL50 mLDiscontinued-Supplier Number: 163371
- 470211-294600 mL600 mLPaquete de 5$708.00Case de 10 (5UN)$1,305.79Supplier Number: 430823