Order Entry
Mexico
Orders LinkContactUsLinkComponent
<i>Cucurbita</i> Stem Combination Slide
Cucurbita Stem Combination Slide
Cucurbita Stem Combination, Cross section & longitudinal section, Quadruple stained
Catalog # 470177-156
Supplier:   Avantor
<i>Cucurbita</i> Stem Combination Slide
Catalog # 470177-156
Supplier:  Avantor
<i>Cucurbita</i> Stem Combination Slide
Cucurbita Stem Combination Slide
Cucurbita Stem Combination, Cross section & longitudinal section, Quadruple stained
Price
$344.40
Log in for availability
Each (1UN)
Product Details & Documents
Illustrates Structure of Gourd Plant Stem
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows both sieve plates and tubes.
Specifications
  • Description:
    Cucurbita Stem Combination
  • Stain:
    Quadruple stained
  • Orientation:
    Cross section & longitudinal section