Order Entry
Mexico
Orders LinkContactUsLinkComponent
Crucible Holder
Crucible Holder
Crucible Holder
Catalog # 470151-210
Supplier #:  VCHWGS
Crucible Holder
Catalog # 470151-210
Crucible Holder
Crucible Holder
Crucible Holder
Price
$319.78
Log in for availability
Each (1UN) Case de 6 (1UN)
Product Details & Documents
Made of molded, black rubber with a conical opening 27 mm at the top tapered to 22 mm at the bottom, the crucible holder includes a glass funnel.
  • Holder may be used with suction flasks of 250 to 1000 ml capacity
  • Holds Gooch and similar crucibles of 35 to 40 mm diameter at the top
Overall length: 60 mm.
Specifications
  • Description:
    Crucible Holder