Cross Colored Threads Slide
Cross Colored Threads, whole mount, Whole mount, Unstained
Catalog # 470176-898
Supplier: Avantor
Jump to:
Product Details & Documents
Whole mount
- Teaches basic microscopy skills and lens characteristics
- Rigorous quality control standards
For teaching students to focus the microscope. Yellow on top, followed by blue, with red on the bottom. Threads intersect at one point.
Specifications
- Description:Cross Colored Threads, whole mount
- Stain:Unstained
- Orientation:Whole mount