Order Entry
Mexico
ContactUsLinkComponent
Corning Microprocessor Controlled Stirrer
Corning Microprocessor Controlled Stirrer
Catalog # 470211-558
Supplier:  CORNING CA
undefined
Corning Microprocessor Controlled Stirrer
Catalog # 470211-558
Supplier:  CORNING CA
Supplier Number:  6795-210
Specifications
  • Description:
    Corning Microprocessor Controlled Stirrer
  • Cat. no.:
    470211-558
  • Supplier No.:
    6795-210
Specifications
About this item
The Corning® stirrers have a durable glass-ceramic 4×5" Pyroceram® top and offer superior performance and reliability in a low profile, space-saving design.
  • Designed with CEE7-7 plug
  • Microprocessor controlled to automatically adjust motor strength to any solution
  • Enclosed electronics design protects components, improves performance, and prolongs product life
  • Two piece casting design deflects spills away from electronics
  • Built in mount for an optional support rod
Exclusive closed-loop stirring control monitors and regulates the stirring speed for aqueous, viscous, or semi-solid solutions. A stronger, larger magnet improves stir bar coupling and stirring capability.
Delivery: Includes a built in mount for an optional support rod and 1×9.5 stir bar. Manufacturer's two-year warranty on parts and labor. Instruction manual, technical information book, and three-wire cord and plug.