Order Entry
Mexico
Orders LinkContactUsLinkComponent
Lab-Aids® Corn Crop Genetics Kit
Lab-Aids® Corn Crop Genetics Kit
Corn Crop Genetics Kit, 32
Catalog # 470012-426
Supplier #:  603S
Lab-Aids® Corn Crop Genetics Kit
Catalog # 470012-426
Lab-Aids® Corn Crop Genetics Kit
Lab-Aids® Corn Crop Genetics Kit
Corn Crop Genetics Kit, 32
Price
$3,431.52
Log in for availability
Each (1UN)
Product Details & Documents
Includes Realistic Corn Ear Cutouts
  • Reusable
  • Uses images of corn ears for consistant results
  • Developed by Sepup
  • Time required: single 50 minute lab
Students build their understanding of selective breeding for desired traits in crops such as corn. They investigate the outcomes of crosses of hybrid and dihybrid corn plants for two unlinked genetic traits: corn kernel color and sweetness. They use Punnett squares to predict the ratio of phenotypes that will be produced by different crosses and analyze the actual ratio of phenotypes of the second generation offspring. A reading describes traditional selective breeding and modern genetic engineering approaches to improving crops to improving crops.
Delivery information: See datasheet for contents information.
Documents
Specifications
  • Description:
    Corn Crop Genetics Kit
  • No. of Lab Groups:
    32