Order Entry
Mexico
ContactUsLinkComponent
Core-Valence-Radius Periodic Table
Core-Valence-Radius Periodic Table
Catalog # 470007-256
undefined
Core-Valence-Radius Periodic Table
Catalog # 470007-256
Supplier Number:  6730921
Specifications
  • Description:
    Core-Valence-Radius Periodic Table
  • Cat. no.:
    470007-256
  • Supplier No.:
    6730921
Specifications
About this item
Developed by Teacher James Ross, the Core-Valence-Radius Periodic Table is a new type of periodic table designed to make learning chemistry a more intuitive process. Each element is represented by a nucleus (core), a valence shell, and an electronegativity value.
  • The table includes rows 1 to 4 and families 1, 2 and 13 to 18 (IA - VIIIA)
  • Size: 91.4×144.8 cm
Use the Core-Valence-Radius Periodic Table in conjunction with table manners by Jim Ross and Mike Lattner. This introductory book covers material from the introduction of the periodic table, metals, non-metals and metalloids, reactivity of elements, gaining and losing electrons, ionic and covalent bonding; combustion and other reactions; some simple acid/base chemistry.
Also available by Jim Ross and Mike Lattner is Table Talk. This book covers redox and oxidation numbers, covalent bonding, dipoles and polarity in small (mostly organic) molecules, acid/base properties of organic and other molecules, and more. Grades 8 to 11.