TeacherGeek Connector Strips, 300mm
Catalog # 470132-430
Supplier: TEACHERGEEK INC
Specifications
- Description:Connector Strips (300 mm) pack of 50
- Cat. no.:470132-430
- Supplier No.:27005
- No. of Parts:50
Specifications
About this item
Bulk connector strip to expand your TeacherGeek collection
- Basic structural components
- Strips can be easily cut
TeacherGeek 300mm Connector Strips are basic structural components to build bridges, vehicle chassis, hydraulic arms, crazy contraptions, launchers, structures or original design innovations. The splined holes securely hold dowels or can be reamed for axle rotation. These strips can be easily cut to your desired size.