Complete Blood Pressure Kit
Catalog # 470019-078
Supplier: HEALTH SMART SE
Specifications
- Description:Complete Blood Pressure Kit
- Cat. no.:470019-078
- Supplier No.:04-176-021
Specifications
About this item
Everything needed to measure blood pressure and pulse is included in this economical kit.
- Accurate to ±3 mm Hg
- Zippered, Vinyl Storage Case
- Included Recording Chart for Individual Record Keeping
It includes a stethoscope and a zippered vinyl storage case with instructions and record chart.
The provided aneroid sphygmomanometer, which features a standard-size D-ring cuff with a Velcro closure, is accurate to ±3 mm Hg.