Comparative Plant Anatomy Slide Set
470183-472
: Avantor
- Type:Plant
- Description:Comparative Plant Anatomy Slide Set
- No. of Slides:6
- Cat. no.:470183-472
- Supplier No.:WARD470183-472
Illustrate Basic Morphologic Divergence Between the Monocot and Dicot Plant Groups
- Identifying features clearly distinguishable
- Rigorous quality control standards
: Set includes Monocot and dicot roots (cs); Primary and secondary stems (cs); Dicot and monocot stems (cs); Dicot and monocot leaves (cs); Air and water leaves (cs); and Leaf types (cs).