Order Entry
Mexico
ContactUsLinkComponent
Colon, Mammal, Mucus Goblet Cells Slide
Colon, Mammal, Mucus Goblet Cells Slide
Catalog # 470182-566
Supplier:  Avantor
undefined
Colon, Mammal, Mucus Goblet Cells Slide
Catalog # 470182-566
Supplier:  Avantor
Specifications
  • Description:
    Colon, Mammal, Mucus Goblet Cells
  • Cat. no.:
    470182-566
  • Stain:
    Periodic acid Schiff reaction
  • Supplier No.:
    WARD470182-566
  • Orientation:
    Cross Section
Specifications
About this item
Illustrates a Segment of the Digestive Tract
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows goblet cells, stained red.