Order Entry
Mexico
Orders LinkContactUsLinkComponent
Colon Slide
Colon Slide
Colon, Cross section, Hematoxylin and Eosin
Catalog # 470177-128
Supplier:   Avantor
Colon Slide
Catalog # 470177-128
Supplier:  Avantor
Colon Slide
Colon Slide
Colon, Cross section, Hematoxylin and Eosin
Price
$222.60
Log in for availability
Each (1UN)
Product Details & Documents
Illustrates a Segment of the Digestive Tract
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Specifications
  • Description:
    Colon
  • Stain:
    Hematoxylin and Eosin
  • Orientation:
    Cross section