Order Entry
Mexico
Orders LinkContactUsLinkComponent
Chromatography Paper
46 x 57 cm (18 x 22"), 130 mm/30 min
  470003-724
 :  6130-4657
Chromatography Paper
  470003-724
Chromatography Paper
Chromatography Paper
46 x 57 cm (18 x 22"), 130 mm/30 min
$2,678.37
Paquete de 100
Quality paper for general chromatography and electrophoresis.
  • White, smooth sheets with a hard surface
  • Flow rate:
    130 mm/30 min
  • Size:
    46 x 57 cm (18 x 22")