Ceramic Fiber Squares
Supplier: ARMIL/CFS INC SE
Jump to:
About this item
Made of ceramic fiber with organic binder, for use under hot vessels to protect table tops. Does not contain asbestos. Package of 12.
1 Options Available Below
Product Details & Documents
Made of ceramic fiber with organic binder, for use under hot vessels to protect table tops. Does not contain asbestos. Package of 12.
Product Family Options
Product Information
- AvailabilityPrice
- 470122-536Ceramic Fiber Squares, 13 x 13 cmSupplier Number: 550F-5X5
- 470122-538Ceramic Fiber Squares, 15 x 15 cmCeramic Fiber Squares, 15 x 15 cmDiscontinued-Supplier Number: 550F-6X6