Order Entry
Mexico
Orders LinkContactUsLinkComponent
Cellular Anatomy Notebook Chart

Cellular Anatomy Notebook Chart

Cellular Anatomy Notebook Chart, Laminated, 28 cm, 20 cm
  470101-368
Supplier #:  1550803778
Cellular Anatomy Notebook Chart
  470101-368
Cellular Anatomy Notebook Chart
Cellular Anatomy Notebook Chart

Cellular Anatomy Notebook Chart, Laminated, 28 cm, 20 cm

 
$174.31
   
Each (1UN)
 

Introduce your students to cellular anatomy with this vivid, full-color, two-sided chart.

  • Laminated for durability

One side depicts the richly detailed, inner structure of a plant cell and organelles, while the other is a generalized animal cell with organelles. The result of extensive research from multiple sources, each component is clearly labeled. The animal cell side features plasma membrane, nuclear envelope, and cytoskeleton details. This quick reference chart is laminated for durability and protection against spills and other damage.

 
  • Width:
    20 cm
  • Description:
    Cellular Anatomy Notebook Chart
  • Length:
    28 cm
  • Finish:
    Laminated
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.