Cartilage, Composite Slide
Cartilage, Composite Slide, Section, Verhoeff and Eosin
Catalog # 470182-642
Supplier: Avantor
Jump to:
Product Details & Documents
Investigate the characteristics of cartilage with and without elastic fibers
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Section of hyaline and elastic cartilage showing the presence and absence of elastic fibers.
Specifications
- Description:Cartilage, Composite Slide
- Stain:Verhoeff and Eosin
- Orientation:Section