Order Entry
Mexico
Orders LinkContactUsLinkComponent
Foam Calorimeter Set
Calorimeter Set, Foam
  470012-650
 :   Avantor
Foam Calorimeter Set
  470012-650
 :  Avantor
Foam Calorimeter Set
Foam Calorimeter Set
Calorimeter Set, Foam
$1,467.13
Each (1UN) Paleta de 60 (1UN)
Ensure your studies of heat exchange are safe, easy, and fun.
These economical calorimeters features slotted lids to allow insertion of a thermometer to take measutremnts without losing heat to the surrounding air. Each holds 500 ml of water.
Delivery information: Includes a set of 10 calorimeters and instructions.
  • Description:
    Calorimeter Set, Foam