V Jaw Buret Clamp
Catalog # 470104-098
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Description:V Jaw Buret Clamp
- Cat. no.:470104-098
- Supplier No.:COBR3
Specifications
About this item
Hold a variety of small labware items from glassware to metal.
- PVC coated jaws
- Jaws can be rotated to any angle
Ideal for holding small objects, this clamp has one flat jaw and one V-shaped jaw, and provides an opening of 5-35 mm. The jaws can be rotated 360° and locked at any angle. The clamp attaches quickly to standard support rods up to 13 mm in diameter.