Order Entry
Mexico
ContactUsLinkComponent
Bow-Type Drawing Compass
Bow-Type Drawing Compass
Catalog # 470122-026
Proveedor:  CHARTPAK INC SE
undefined
Bow-Type Drawing Compass
Catalog # 470122-026
Proveedor:  CHARTPAK INC SE
Supplier Number:  401N

Especificaciones

  • Description:
    Bow-type drawing compass
  • Cat. no.:
    470122-026
  • Supplier No.:
    401N

Especificaciones

Acerca de este artículo

Chrome plated, 6" bow pencil compass. Adjustable for circles up to 10".