Order Entry
Mexico
Orders LinkContactUsLinkComponent
Human Bone, Ground Preparation Slide
Human Bone, Ground Preparation Slide
Bone (Human), Ground Preparation, Cross Section, Unstained
Catalog # 470177-628
Supplier:   Avantor
Human Bone, Ground Preparation Slide
Catalog # 470177-628
Supplier:  Avantor
Human Bone, Ground Preparation Slide
Human Bone, Ground Preparation Slide
Bone (Human), Ground Preparation, Cross Section, Unstained
Price
$193.60
Log in for availability
Each (1UN)
Product Details & Documents
cross section, unstained, thin section
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Thin section showing Haversian canals, lacunae, and canaliculi.
Specifications
  • Description:
    Bone (Human), Ground Preparation
  • Stain:
    Unstained
  • Orientation:
    Cross Section