Bel-Bulb Pipettor
Bel-Bulb Pipettor, Pack of 2
Catalog # 470145-138
Supplier: BEL-ART PRODUCTS SE
Supplier #: F37887-0000_SE
Jump to:
Product Details & Documents
A time saver that eliminates mouth pipetting!
The Bel-bulb Pipettor is simply held against the pipette end and fluid is drawn up.
When the pipette is full, the bulb is removed and the pipette end is covered with the finger to hold the fluid or the fluid can be released as necessary.
Designed to fit pipettes from 6 to 8 mm in Ø, the flexible vinyl bulb is 51 mm in Ø by 70 mm long with a soft tapered chuck.
Specifications
- Description:Bel-Bulb Pipettor, Pack of 2