Beaker Starter Set
50, 100, 250, 600, 1000 ml, Beaker Starter Set
Catalog # 470122-762
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: BGSET5
Jump to:
Product Details & Documents
An ideal starter set for any laboratory.
- Feature pour spouts
- Double-graduated metric scale and durable marking area
A set of 5 beakers of the most commonly used sizes.
: Contains one each of the 50, 100, 250, 600, and 1000 ml beakers.
Specifications
- Capacity:50, 100, 250, 600, 1000 ml
- Description:Beaker Starter Set