Order Entry
Mexico
Orders LinkContactUsLinkComponent
Barr Bodies Present Slide
Barr Bodies Present Slide
Barr Bodies Present, Smear, Biebrich and Fast Green
Catalog # 470183-140
Supplier:   Avantor
Barr Bodies Present Slide
Catalog # 470183-140
Supplier:  Avantor
Barr Bodies Present Slide
Barr Bodies Present Slide
Barr Bodies Present, Smear, Biebrich and Fast Green
Price
$270.90
Log in for availability
Each (1UN)
Product Details & Documents
Illustrates Genetic "Femaleness" in Humans
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Smear of isolated squamous epithelial cells from the buccal cavity of a normal human female. Many nuclei show Barr bodies or drumstick chromosomes.
Specifications
  • Description:
    Barr Bodies Present
  • Stain:
    Biebrich and Fast Green
  • Orientation:
    Smear