Barr Bodies, Male/Female Comparison Slide
Catalog # 470183-144
Supplier: Avantor
Specifications
- Description:Barr Bodies, Male/Female Comparison
- Cat. no.:470183-144
- Stain:Biebrich and Fast Green
- Supplier No.:WARD470183-144
- Orientation:Smear
Specifications
About this item
Illustrates Genetic "Femaleness" in Humans
- Identifying features clearly distinguishable
- Rigorous quality control standards
Scrapings from both human male and female mouth cavities on a single slide to demonstrate the presence of Barr bodies in the female cells and their absence in the male cells.