Bare Copper Wire, 454 g Spool
Wire, Bare Copper, 24 Gauge, 250 m, 454 grams
Catalog # 470201-538
Supplier: ARCOR ELECTRONICS CO INC SE
Supplier #: F-24
Jump to:
Product Details & Documents
A bare copper wire of the highest quality. Available in several standard AWG sizes.
- 454 g spool
Specifications
- Description:Wire, Bare Copper, 24 Gauge, 250 m, 454 grams
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



