Bacillus cereus var. mycoides Slide
Catalog # 470176-832
Supplier: Avantor
Specifications
- Description:Bacillus cereus var. mycoides
- Cat. no.:470176-832
- Stain:Gram (+)
- Supplier No.:WARD470176-832
- Orientation:Smear
Specifications
About this item
Food Poisoning Agent
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods occurring in chains. Endospores formed.