Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Augite
 :   Avantor
Ward's® Augite
 :  Avantor
Ward's® Augite
Ward's® Augite
Pyroxene group; dark green, pure cleaved fragments.
  • Collected in Quebec, Canada
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Description
    Specimen Dimensions
  • Augite, hand specimen, each
    2×3 - 3×4"
    Each (1UN)
    Each (1UN) - $335.65
    $335.65
    Each (1UN)
       $335.65
  • Augite, student specimens
    1×1 - 1×1"
    Paquete de 10
    Paquete de 10 - $416.38
    $416.38
    Paquete de 10
       $416.38
  • Augite, bulk pack, 1 kg
    Size will vary
    Each (1UN)
    Each (1UN) - $512.05
    $512.05
    Each (1UN)
       $512.05
  • Augite thin section
    Each (1UN)
    Each (1UN) - $1,045.20
    $1,045.20
    Each (1UN)
       $1,045.20