Order Entry
Mexico
Orders LinkContactUsLinkComponent
Artery Slide
Artery Slide
Artery Slide, Cross section, Verhoeff and Eosin
Catalog # 470183-008
Supplier:   Avantor
Artery Slide
Catalog # 470183-008
Supplier:  Avantor
Artery Slide
Artery Slide
Artery Slide, Cross section, Verhoeff and Eosin
Price
$135.18
Log in for availability
Each (1UN)
Product Details & Documents
Note the elastic tissue within the artery.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Medium size. Elastic tissue stained.
Specifications
  • Description:
    Artery Slide
  • Stain:
    Verhoeff and Eosin
  • Orientation:
    Cross section