Order Entry
Mexico
ContactUsLinkComponent
CENCO® AP Physics Lab 15: Temperature of a Bunsen Burner Flame
  470018-138
 :  Avantor
undefined
CENCO® AP Physics Lab 15: Temperature of a Bunsen Burner Flame
  470018-138
 :  Avantor
  • Description:
    CENCO AP Physics Lab 15: Temperature of a Bunsen Burner Flame
  • Environmentally Preferable:
  • Cat. no.:
    470018-138
  • Supplier No.:
    WARD470018-138
Practice Calorimetry Techniques
  • Durable materials for years of use
  • Instruction manual included
Introducing the new Cenco AP Physics lab series. Ideal for AP, B and C, I, II, and advanced honors physics courses, each of the 24 experiments is aligned with the College Board AP Curriculum. The teacher guides and student manuals are authored by an experienced AP Physics teacher, so you can trust the setups to be successful in your classroom. Lead students through these hands-on labs using the outlined guided-inquiry approach. All labs come complete with instructions and materials for up to six students per lab group, the inquiry based instructions are found in the separate manuals. For your convenience, labs are packaged to allow you to purchase only what is required for each experiment.
Students use calorimetry to determine the temperature of a bunsen burner flame in this advanced level lab. A large steel washer is held in the flame of a bunsen burner and allowed to come to thermal equilibrium. Students then use calorimetry techniques to determine the flame's temperature. Students will also learn about proper lab usage for a burner. The kit includes two 1" steel nuts, matches, 10 gauge copper wire, hooded lighter, crucible tongs, six insulated cups, and a digital thermometer. Required but not included are a bunsen burner and support stands.
*AP and the Advanced Placement Program are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.