Order Entry
Mexico
ContactUsLinkComponent
Algal Dolomite (Silurian)
Algal Dolomite (Silurian)
  470027-504
 :  Avantor
undefined
Algal Dolomite (Silurian)
  470027-504
 :  Avantor
  • Description:
    Algal Dolomite
  • Cat. no.:
    470027-504
  • Supplier No.:
    WARD470027-504
Sawn and lacquered slabs showing finely banded stromatolitic structures.
  • Cyanobacteria
  • Silurian
Average size: 2.5×2.5×1" to 3×4×1".