Order Entry
Mexico
Orders LinkContactUsLinkComponent
Pulley
Pulley
Single sheave
Catalog # 470049-642
Supplier #:  PULS01
Pulley
Catalog # 470049-642
Pulley
Pulley
Single sheave
Price
$32.53
Log in for availability
Each (1UN)
Product Details & Documents
Smooth running pulley for investigating mechanical advantage.
  • V grooves
  • Swivel hooks
These free-running, sturdy pulleys feature 50mm diameter plastic sheaves with deep V-grooves, aluminum frames, and swivel hooks on both ends.
Specifications
  • Description:
    Single sheave