Clamp Pulley, Adjustable
Clamp Pulley, Adjustable
Catalog # 470148-812
Supplier: VWR International
Jump to:
Product Details & Documents
This versatile pulley can be used on table edges or at different heights.
- Nylon Construction
- Steel Cone Bearing
- Rod Length: 15 cm
- Pulley Works on Table Edges or at Different Heights
- For Experiments Requiring Pulleys to Have Support at Various Heights
It's ideal for experiments that require a pulley to be supported at various heights.
The 40 mm nylon pulley is mounted on steel cone bearings in a metal fork. The fork is attached to a 15 cm rod.
Specifications
- Description:Clamp Pulley, Adjustable
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



