Order Entry
Mexico
Orders LinkContactUsLinkComponent
Acrylic Ray Optics Set
Acrylic Ray Optics Set
  470149-474
Acrylic Ray Optics Set
  470149-474
Acrylic Ray Optics Set
Acrylic Ray Optics Set
Acrylic Ray Optics Set
$685.24
Each (1UN) Case de 10 (1UN)
A Good Optics Set for Demonstrations or First Experiments
  • Made of polished acrylic
  • Provided in a convenient storage box
Set consists of: rectangle (75mm slides), double convex lens and two double concave lenses with a focal length of about 10cm, equilateral prism, and semi-circular block. 15mm thick.
  • Description:
    Acrylic Ray Optics Set