GPI Anatomicals® 4-Stage Osteoporosis Disk Set
470126-648
Small Business Enterprise
- Description:4-Stage Osteoporosis Disk Set
- Height:4 cm
- Length:10 cm
- No. of Keyed Items:4
- Cat. no.:470126-648
- Width:10 cm
- Supplier No.:1650
- No. of Parts:1
Enlarged Cross–Sectional Depiction
Illustrate the progressive thinning of trabeculae bone that occurs due to osteoporosis with these enlarged cross-sections of bone. Four stages of osteoporosis are illustrated on hinged discs that conveniently fold into one another for compact storage and easy portability. This model makes an ideal teaching aid for introductory lessons, as well as a useful professional tool for providing patients with detailed explanations of the disease.