Order Entry
Mexico
ContactUsLinkComponent
Ethanol absolute (denatured with 5% methanol) ≥95% for biochemistry
Flammable
Toxic
Health
Ethanol absolute (denatured with 5% methanol) ≥95% for biochemistry
Catalog # 95043-302
Supplier:  IBI SCIENTIFIC
CAS Number:  
undefined
Flammable
Toxic
Health
Ethanol absolute (denatured with 5% methanol) ≥95% for biochemistry
Catalog # 95043-302
Supplier:  IBI SCIENTIFIC
Supplier Number:  IB15724
CAS Number:  

Specifications

  • Size:
    4 L
  • Cat. no.:
    95043-302
  • Supplier No.:
    IB15724

Specifications

Specification Test Results

  • Purity:
    95%
  • DNase assay:
    None Detected
  • RNase assay:
    None Detected
  • Methanol :
    5%
  • Moisture (KF):
    1% Max.

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:∼78 °C (1013 hPa)
Boiling Pt:~ 78 °C (1013 hPa)
Density:0.8 g/cm³ (20 °C)
Flash Pt:13 °C (55°F)
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568