Round Winpaks®, High-Density Polyethylene, Qorpak®
Catalog # 11311-368
Supplier: QORPAK
Small Business Enterprise
Specifications
- Cap Size:70-400
- Capacity:19 L (5 gal.)
- Color:Natural
- Description:Natural Polyethylene Gasket Lined
- Material:HDPE
- Cat. no.:11311-368
- Supplier No.:PLA-03327
Specifications
About this item
Winpaks® are cylindrical in shape with spout placement in line with the sidewall for maximum removal of product during dispensing.
The unit is also equipped with a swing handle on the top face. Designed for the secure, dependable shipment and storage of liquid products.