Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for sensION+ pH Electrodes, Hach
Accessories for sensION+ pH Electrodes, Hach
89174-438
:
Avantor
Product available on GSA Advantage®
Accessories for sensION+ pH Electrodes, Hach
89174-438
:
Avantor
Product available on GSA Advantage®
Description:
Electrolytic solution (KCl 3M), 125mL flask sensION+
Cat. no.:
89174-438
Supplier No.:
LZW9510.99