Potassium hydrogen phthalate
Poly bottle, 100 g
Catalog # 220202-348
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
Supplier #: P1088-100G
Potassium hydrogen phthalate
Catalog # 220202-348
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
CAS Number: 877-24-7
Jump to:
Product Details & Documents
Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
- CAS Number:877-24-7
- Formula:HOOCC₆H₄COOK
- MDL Number:MFCD00013070
- Molecular Weight:204.22 g/mol
- Merck Index:13,07694
- Melting Pt:295 - 300 °C
- Melting Pt:295…300 °C
- Density:1.6362 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:212-889-4
Documents
Specifications
Specifications
- Pack type:Poly bottle
- Size:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



