Dimidium bromide
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
Jump to:
About this item
Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
- CAS Number:518-67-2
- Formula:C₂₀H₁₈BrN₃
- MDL Number:MFCD00011757
- Molecular Weight:380.29 g/mol
- Melting Pt:243 - 248 °C
- Melting Pt:243…248 °C
- Storage Temperature:Ambient
- EINECS:208-256-7
- UN:2811
- ADR:6.1,I
2 Options Available Below
Product Details & Documents
Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
- CAS Number:518-67-2
- Formula:C₂₀H₁₈BrN₃
- MDL Number:MFCD00011757
- Molecular Weight:380.29 g/mol
- Melting Pt:243 - 248 °C
- Melting Pt:243…248 °C
- Storage Temperature:Ambient
- EINECS:208-256-7
- UN:2811
- ADR:6.1,I
Documents
Sigma Product Details
Additional Product Details
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



