Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Ethyl formate ≥97%, Reagent Grade

Bottle, 50 ml
Catalog # 220033-626
Flammable
Irritant
 :  112682-500ML
Ethyl formate ≥97%, Reagent Grade
Catalog # 220033-626
CAS Number:  109-94-4
Ethyl formate ≥97%, Reagent Grade

Bottle, 50 ml

CAS Number:  109-94-4
Price
$2,552.04
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Formic acid ethyl ester
  • CAS Number:109-94-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00003294
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,03841
  • Boiling Pt:148 °C (1013 hPa)
  • Melting Pt:-2 °C
  • Melting Pt:–2 °C
  • Density:0.9208 g/cm³ (20 °C)
  • Flash Pt:-20 °C
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • EINECS:203-721-0
  • UN:1190
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    50 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.