Order Entry
Mexico
ContactUsLinkComponent
Carbon disulphide ≥99.9% CHROMASOLV™ for HPLC
Carbon disulphide ≥99.9% CHROMASOLV™ for HPLC
Catalog # 220050-124
CAS Number:  
undefined
Carbon disulphide ≥99.9% CHROMASOLV™ for HPLC
Catalog # 220050-124
Supplier Number:  270660-2L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    2 L
  • Cat. no.:
    220050-124
  • Supplier No.:
    270660-2L

Specifications

About this item

 :
 :76.14 g/mol
Merck Index:13,01821
Boiling Pt:46.2 °C (1013 hPa)
Boiling Pt:46.2 °C (1013 hPa)
Melting Pt:–111.6 °C
Melting Pt:-111.6 °C
Density:1.2632 g/cm³ (20 °C)
Flash Pt:–30 °C
Flash Pt:-30 °C
EINECS:200-843-6
UN:1131
ADR:3,I
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321