Hydroxylammonium chloride, ReagentPlus® analytical reagent
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
Hydroxylammonium chloride, ReagentPlus® analytical reagent
Supplier: SIGMA ALDRICH QUIMICA,S DE RL MX
Certificates
Jump to:
About this item
Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:151 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-798-2
- UN:2923
- ADR:8,III
2 Options Available Below
Product Details & Documents
Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:151 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



