Order Entry
Mexico
ContactUsLinkComponent
 
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Catalog # BDH1160-4LP
Supplier:  VWR International
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Catalog # BDH1160-4LP
Supplier:  VWR International
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    4 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
  • Cat. no.:
    BDH1160-4LP
  • Supplier No.:
    PL092-4P

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless liquid
  • Water:
    9.0 – 11.0%
  • Residue:
    Less than 0.001% (0.0010g/124mL)
  • GC Purity (Ethanol):
    81.0-82.0%
  • GC Purity (Methanol):
    3.8-4.5%
  • GC Purity (Isopropyl Alcohol):
    4.0-5.0%
  • Color (APHA):
    Less than 10
  • Specific gravity @ 20ºC:
    0.822-0.829
  • Acidity:
    Less than 0.0025%

Specification Test Results

About this item

Synonyms: Ethyl alcohol denatured
 :
 :46.07 g/mol
Boiling Pt:78.3 °C (1013 hPa)
Flash Pt:12 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568