KleenGuard™ V80 Revolution™ Protective Goggles, Kimberly-Clark Professional
Catálogo # 89135-145
Supplier: Ansell Healthcare
Specifications
- Frame Color:Blue
- Description:Revolution™ protective goggles
- Cat. no.:89135-145
- Supplier No.:14399
Specifications
About this item
When you need splash protection, anti-fog lenses, and a sleek look all rolled into one, trust KleenGuard™ Revolution™ OTG Safety Goggles.
- Universal size (one size fits most)
- Meets ANSI Z87.1+ standard for impact protection with a D3 rating for droplet and splash protection
- Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
- Indirect ventilation protects from splashes and provides increased airflow to minimize condensation and fogging
- The lens is hardcoated for scratch protection and longer life
- Fits over most prescription eyeglasses and features adjustable headband buckles for a snug fit
KleenGuard™ Revolution™ OTG Safety Goggles feature a soft, pliable frame gasket for enhanced fit and comfort, and cross-vent technology to keep air flowing and moisture out to prevent fogging. The clear lenses allow maximum light transmission and feature an anti-fog coating. They are made of lightweight polycarbonate material that provides 99.9% UVA/UVB/UVC protection, and are hardcoated for scratch resistance and longer life.
Certifications: These safety goggles meet the ANSI Z87.1+ standards for impact, with a D3 rating for droplet and splash protection, and are TAA compliant.