Puritan® Standard Polyester Tipped Applicator, Polystyrene Handle, Sterile, Puritan Medical Products
Supplier: PURITAN MEDICAL PRODUCTS COMP
Certificates
About this item
These swabs have a wide range of applications, including specimen collection for DNA testing, throat swabs, gynecological screenings, and testing industrial surfaces
The standard transport tube is a reclosable dry transport tube ideal for transporting specimens for lab work. The molded transport tube is also reclosable, and features a molded base to hold the tip in center of tube to help avoid loss of sample material. Made in the USA.
The polyester tips are bonded to a semi-flexible polystyrene handle with an aqueous based adhesive
Certifications: CE marked.
Frequently Bought Together