Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ethidium bromide for biotechnology

Ethidium bromide for biotechnology

5 g
Catalog # 97064-602
Toxic
Health
Supplier:   VWR
Número de proveedor #:  0492-5G
Ethidium bromide for biotechnology
Catalog # 97064-602
Supplier:  VWR
CAS Number:  1239-45-8
Ethidium bromide for biotechnology
Ethidium bromide for biotechnology

5 g

CAS Number:  1239-45-8
Price
$4,723.84
Log in for availability
Each (5g) Case de 40 (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

At a concentration of 0.5μg/ml, ethidium bromide is useful in agarose or acrylamide gels for band detection. It is used to detect as little as 10ng of nucleic acid. At higher concentrations (0.5–1.0μg/ml), it may be used to facilitate purification of DNA in a cesium chloride gradient.

  • Fluorescent stain for visualizing nucleic acids

For maximum convenience, AMRESCO offers ethidium bromide in various forms, including ready-to-use solutions and powdered forms. Each is packaged in shatter-resistant packaging for added safety. AMRESCO’s ethidium bromide dropper bottle offers the ultimate convenience and safety. Simply dispense one drop of ethidium bromide into 50ml of agarose gel or running buffer (TBE or TAE) or staining solution.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 -rr)
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261 - 264 °C
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:≤ 100 °C
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I

Documents

Specifications

Specification Test Results

  • DNase:
    none detected
  • Loss on Drying:
    = 5 %
  • Protease:
    none detected
  • Purity (Dry Basis):
    = 98 %
  • RNase:
    none

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.