
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Proveedor: VWR
Certificados
Acerca de este artículo
Número CAS:77-86-1
:
Fórmula:NH₂C(CH₂OH)₃
Número MDL:MFCD00004679
:121.14 g/mol
Índice Merck:13,09842
Punto de ebullición:288°C
Punto de fusión:169°C
Densidad:1.328 g/cm³ (20 °C)
EINECS:201-064-4
:GHS07
Temperatura de almacenamiento:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Resultados de prueba de especificación
- Abs.@280nm (1M, Water):= 0.05
- Arsenic:0.0005 %
- Copper:0.0001 %
- DNase:none detected
- Insolubles:= 0.005 %
- Iron:0.0001 %
- Lead:0.0001 %
- Loss on Drying:1 %
- Magnesium:0.0005 %
- Protease:none
- Purity:99.8 %
- RNase:none detected
Resultados de prueba de especificación
Frequently Bought Together